Anti-UBA2 Rabbit Polyclonal Antibody
Lieferant: Biorbyt
BIRBORB126468-100EA 0 EUR
BIRBORB126468-100 BIRBORB126468-200
Anti-UBA2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: UBA2 (ubiquitin-like modifier activating enzyme 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human