Anti-USP36 Rabbit Polyclonal Antibody
Lieferant: Biorbyt
BIRBORB126478-100EA 0 EUR
BIRBORB126478-100 BIRBORB126478-200
Anti-USP36 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: USP36 (ubiquitin specific peptidase 36)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human