Anti-PARP2 Rabbit Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB127129-100EA 0 EUR
BIRBORB127129-100 BIRBORB127129-200
Anti-PARP2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PARP2 (poly (ADP-ribose) polymerase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR