Anti-PAX5 Rabbit Polyclonal Antibody
Lieferant: Biorbyt
BIRBORB127130-100EA 0 EUR
BIRBORB127130-100 BIRBORB127130-200
Anti-PAX5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PAX5 (paired box 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen