Anti-PFKFB2 Rabbit Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB127134-200EA 0 EUR
BIRBORB127134-200 BIRBORB127134-100
Anti-PFKFB2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PFKFB2 (6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR