Anti-PIGH Rabbit Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB127139-200EA 0 EUR
BIRBORB127139-200 BIRBORB127139-100
Anti-PIGH Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PIGH (phosphatidylinositol glycan anchor biosynthesis, class H)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR