Anti-POLR3E Rabbit Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB127151-200EA 0 EUR
BIRBORB127151-200 BIRBORB127151-100
Anti-POLR3E Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: POLR3E (polymerase (RNA) III (DNA directed) polypeptide E (80kD))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR