Anti-PPRC1 Rabbit Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB127159-200EA 0 EUR
BIRBORB127159-200 BIRBORB127159-100
Anti-PPRC1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PPRC1 (peroxisome proliferator-activated receptor gamma, coactivator-related 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR