Anti-PRPF39 Rabbit Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB127164-200EA 0 EUR
BIRBORB127164-200 BIRBORB127164-100
Anti-PRPF39 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PRPF39 (pre-mRNA processing factor 39)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR