Anti-KCNA3 Rabbit Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB87792-50EA 0 EUR
BIRBORB87792-50
Anti-KCNA3 Rabbit Polyclonal Antibody
Antikörper
Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: KCNA3 (potassium voltage-gated channel, shaker-related subfamily, member 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR