Anti-HIST2H2BE Rabbit Polyclonal Antibody
Lieferant: Biorbyt
BIRBORB87921-50EA 0 EUR
BIRBORB87921-50
Anti-HIST2H2BE Rabbit Polyclonal Antibody
Antikörper
Immunochemistry: 5 ?g/ml; Western Blot: 2 ?g/ml
Type: Primary
Antigen: HIST2H2BE (histone cluster 2, H2be)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen