Anti-HIST4H4 Rabbit polyclonal antibody unconjugated
Lieferant: Biorbyt
BIRBORB87922-50EA 0 EUR
BIRBORB87922-50
Anti-HIST4H4 Rabbit polyclonal antibody unconjugated
Antikörper
Immunochemistry: 5 ?g/ml; Western Blot: 2 ?g/ml
Type: Primary
Antigen: HIST4H4 (histone cluster 4, H4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human