Anti-HSPA5 Rabbit Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB87924-50EA 0 EUR
BIRBORB87924-50
Anti-HSPA5 Rabbit Polyclonal Antibody
Antikörper
Immunochemistry: 10 ?g/ml; Western Blot: 1-3 ?g/ml

Type: Primary
Antigen: HSPA5 (heat shock 70kDa protein 5 (glucose-regulated protein, 78kDa))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR