Anti-HTR1D Rabbit Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB87927-50EA 0 EUR
BIRBORB87927-50
Anti-HTR1D Rabbit Polyclonal Antibody
Antikörper
Immunochemistry: 10 ?g/ml; Western Blot: 2 ?g/ml

Type: Primary
Antigen: HTR1D (5-hydroxytryptamine (serotonin) receptor 1D, G protein-coupled)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR