Anti-Psoriasin Mouse Monoclonal Antibody
Lieferant: Biorbyt
BIRBORB87942-50EA 0 EUR
BIRBORB87942-50
Anti-Psoriasin Mouse Monoclonal Antibody
Antikörper
Immunochemistry: 10 ?g/ml; Western Blot: 1 ?g/ml
Type: Primary
Antigen: Psoriasin
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human