Anti-SMAD6 Rabbit Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB87951-50EA 0 EUR
BIRBORB87951-50
Anti-SMAD6 Rabbit Polyclonal Antibody
Antikörper
Immunochemistry: 5 ?g/ml; Western Blot: 5-2 ?g/ml

Type: Primary
Antigen: SMAD6 (SMAD family member 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR