Anti-TARBP2P Rabbit Polyclonal Antibody
Lieferant: Biorbyt
BIRBORB87955-50EA 0 EUR
BIRBORB87955-50
Anti-TARBP2P Rabbit Polyclonal Antibody
Antikörper
Immunochemistry: 10 ?g/ml
Type: Primary
Antigen: TARBP2P (TAR (HIV-1) RNA binding protein 2 pseudogene)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen