Anti-TNFRSF10A Mouse Monoclonal Antibody

Lieferant: Biorbyt
BIRBORB87961-50EA 0 EUR
BIRBORB87961-50
Anti-TNFRSF10A Mouse Monoclonal Antibody
Antikörper
Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: TNFRSF10A (tumor necrosis factor receptor superfamily, member 10a)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR