Anti-TNFRSF11A Mouse Monoclonal Antibody

Lieferant: Biorbyt
BIRBORB87962-50EA 0 EUR
BIRBORB87962-50
Anti-TNFRSF11A Mouse Monoclonal Antibody
Antikörper
Immunochemistry: 10 ?g/ml; Western Blot: 1-2 ?g/ml

Type: Primary
Antigen: TNFRSF11A (tumor necrosis factor receptor superfamily, member 11a, NFKB activator)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR