Anti-TP73 Rabbit Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB87969-50EA 0 EUR
BIRBORB87969-50
Anti-TP73 Rabbit Polyclonal Antibody
Antikörper
Immunochemistry: 5 ?g/ml; Western Blot: 1-3 ?g/ml

Type: Primary
Antigen: TP73 (tumor protein p73)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR