Anti-UBA2 Rabbit Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB87976-50EA 0 EUR
BIRBORB87976-50
Anti-UBA2 Rabbit Polyclonal Antibody
Antikörper
Immunochemistry: 5 ?g/ml; Western Blot: 5-2 ?g/ml

Type: Primary
Antigen: UBA2 (ubiquitin-like modifier activating enzyme 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR