Anti-HAVCR1 Rabbit Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB87981-50EA 0 EUR
BIRBORB87981-50
Anti-HAVCR1 Rabbit Polyclonal Antibody
Antikörper
Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: HAVCR1 (hepatitis A virus cellular receptor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR