Anti-CD59 Mouse Monoclonal Antibody

Lieferant: Biorbyt
BIRBORB87983-50EA 0 EUR
BIRBORB87983-50
Anti-CD59 Mouse Monoclonal Antibody
Antikörper
Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: CD59 (CD59 molecule, complement regulatory protein)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR