Anti-TPM3 Mouse Monoclonal Antibody

Lieferant: Biorbyt
BIRBORB96311-50EA 0 EUR
BIRBORB96311-50
Anti-TPM3 Mouse Monoclonal Antibody
Antikörper
Immunochemistry: 20 ?g/ml

Type: Primary
Antigen: TPM3 (tropomyosin 3)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR