Anti-ERN1 Rabbit Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB96314-50EA 0 EUR
BIRBORB96314-50
Anti-ERN1 Rabbit Polyclonal Antibody
Antikörper
Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: ERN1 (endoplasmic reticulum to nucleus signaling 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR