Anti-SS18 Rabbit Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB96315-50EA 0 EUR
BIRBORB96315-50
Anti-SS18 Rabbit Polyclonal Antibody
Antikörper
ELISA: 1:1000; Immunochemistry: 5 ?g/ml; Western Blot: 1:500-1:1000

Type: Primary
Antigen: SS18 (synovial sarcoma translocation, chromosome 18)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR