Anti-B2M Mouse Monoclonal Antibody

Lieferant: Biorbyt
BIRBORB96335-50EA 0 EUR
BIRBORB96335-50
Anti-B2M Mouse Monoclonal Antibody
Antikörper
  • Flow Cytometry: 1 mg/ml; Immunochemistry: 1:50; Western Blot: 2-4 mg/ml

Type: Primary
Antigen: B2M (beta-2-microglobulin)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR