Anti-UB Mouse Monoclonal Antibody

Lieferant: Biorbyt
BIRBORB96352-50EA 0 EUR
BIRBORB96352-50
Anti-UB Mouse Monoclonal Antibody
Antikörper
Immunochemistry: 10 ?g/ml; Western Blot: 1:1000

Type: Primary
Antigen: UB (ubiquitin)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR