Anti-CD1C Mouse Monoclonal Antibody [clone: M241]

Lieferant: Abcam
ABCAAB18216-50EA 877 EUR
ABCAAB18216-50
Anti-CD1C Mouse Monoclonal Antibody [clone: M241]
Antikörper
Type: Primary
Antigen: CD1C (CD1c molecule)
Clonality: monoclonal
Clone: M241
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR