Anti-CDC20 Mouse Monoclonal Antibody [clone: BA8]
Lieferant: Abcam
ABCAAB18217-100EA 903 EUR
ABCAAB18217-100
Anti-CDC20 Mouse Monoclonal Antibody [clone: BA8]
Antikörper
Type: Primary
Antigen: CDC20 (cell division cycle 20)
Clonality: monoclonal
Clone: BA8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestellen