Anti-CCNB1 Mouse monoclonal antibody unconjugated [clone: V92.1]
Lieferant: Abcam
ABCAAB18221-50EA 913 EUR
ABCAAB18221-50
Anti-CCNB1 Mouse monoclonal antibody unconjugated [clone: V92.1]
Antikörper
Type: Primary
Antigen: CCNB1 (cyclin B1)
Clonality: monoclonal
Clone: V92.1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse