Anti-CD34 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 4H11[APG]]

Lieferant: Abcam
ABCAAB18227-50EA 445 EUR
ABCAAB18227-50
Anti-CD34 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 4H11[APG]]
Antikörper
Type: Primary
Antigen: CD34 (CD34 molecule)
Clonality: monoclonal
Clone: 4H11[APG]
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR