Anti-DDDDK tag Mouse monoclonal antibody unconjugated [clone: F-tag-01]
Lieferant: Abcam
ABCAAB18230-100EA 941 EUR
ABCAAB18230-100
Anti-DDDDK tag Mouse monoclonal antibody unconjugated [clone: F-tag-01]
Antikörper
Type: Primary
Antigen: DDDDK tag (DDDDK tag)
Clonality: monoclonal
Clone: F-tag-01
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen