Anti-PXN Rabbit Monoclonal Antibody [clone: Y113]
Lieferant: Abcam
ABCAAB32084-100EA 993 EUR
ABCAAB32084-100
Anti-PXN Rabbit Monoclonal Antibody [clone: Y113]
Antikörper
Type: Primary
Antigen: PXN (paxillin)
Clonality: monoclonal
Clone: Y113
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen