Anti-EGFR Rabbit monoclonal antibody unconjugated [clone: Y39]

Lieferant: Abcam
ABCAAB32086-100EA 697 EUR
ABCAAB32086-100
Anti-EGFR Rabbit monoclonal antibody unconjugated [clone: Y39]
Antikörper
Type: Primary
Antigen: EGFR (epidermal growth factor receptor)
Clonality: monoclonal
Clone: Y39
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR