Anti-PI 3 Kinase catalytic subunit gamma Rabbit Monoclonal Antibody [clone: Y388]

Lieferant: Abcam
ABCAAB32089-100EA 896 EUR
ABCAAB32089-100
Anti-PI 3 Kinase catalytic subunit gamma Rabbit Monoclonal Antibody [clone: Y388]
Antikörper
Type: Primary
Antigen: PI 3 Kinase catalytic subunit gamma
Clonality: monoclonal
Clone: Y388
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR