Anti-CREB Rabbit monoclonal antibody unconjugated [clone: E113]
Lieferant: Abcam
ABCAAB32096-100EA 998 EUR
ABCAAB32096-100
Anti-CREB Rabbit monoclonal antibody unconjugated [clone: E113]
Antikörper
Type: Primary
Antigen: CREB
Clonality: monoclonal
Clone: E113
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen