Anti-ARRB1 Rabbit Monoclonal Antibody [clone: E274]

Lieferant: Abcam
ABCAAB32099-100EA 763 EUR
ABCAAB32099-100
Anti-ARRB1 Rabbit Monoclonal Antibody [clone: E274]
Antikörper
Type: Primary
Antigen: ARRB1 (arrestin, beta 1)
Clonality: monoclonal
Clone: E274
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR