Anti-JAK2 Rabbit Monoclonal Antibody [clone: E132]

Lieferant: Abcam
ABCAAB32101-100EA 779 EUR
ABCAAB32101-100
Anti-JAK2 Rabbit Monoclonal Antibody [clone: E132]
Antikörper
Type: Primary
Antigen: JAK2 (Janus kinase 2)
Clonality: monoclonal
Clone: E132
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR