Anti-PPP2R1A Rabbit Monoclonal Antibody [clone: E155]

Lieferant: Abcam
ABCAAB32104-100EA 749 EUR
ABCAAB32104-100
Anti-PPP2R1A Rabbit Monoclonal Antibody [clone: E155]
Antikörper
Type: Primary
Antigen: PPP2R1A (protein phosphatase 2, regulatory subunit A, alpha)
Clonality: monoclonal
Clone: E155
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR