Anti-ELK1 Rabbit monoclonal antibody unconjugated [clone: Y77]

Lieferant: Abcam
ABCAAB32106-100EA 688 EUR
ABCAAB32106-100
Anti-ELK1 Rabbit monoclonal antibody unconjugated [clone: Y77]
Antikörper
Type: Primary
Antigen: ELK1 (ELK1, member of ETS oncogene family)
Clonality: monoclonal
Clone: Y77
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR