Anti-GRB2 Rabbit Monoclonal Antibody [clone: Y301]

Lieferant: Abcam
ABCAAB32111-100EA 688 EUR
ABCAAB32111-100
Anti-GRB2 Rabbit Monoclonal Antibody [clone: Y301]
Antikörper
Type: Primary
Antigen: GRB2 (growth factor receptor-bound protein 2)
Clonality: monoclonal
Clone: Y301
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR