Anti-RPS6KA1 Rabbit Monoclonal Antibody [clone: E4]

Lieferant: Abcam
ABCAAB32114-100EA 1020 EUR
ABCAAB32114-100
Anti-RPS6KA1 Rabbit Monoclonal Antibody [clone: E4]
Antikörper
Type: Primary
Antigen: RPS6KA1 (ribosomal protein S6 kinase, 90kDa, polypeptide 1)
Clonality: monoclonal
Clone: E4
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR