Anti-PXN Rabbit Monoclonal Antibody [clone: E228]
Lieferant: Abcam
ABCAAB32115-100EA 913 EUR
ABCAAB32115-100
Anti-PXN Rabbit Monoclonal Antibody [clone: E228]
Antikörper
Type: Primary
Antigen: PXN (paxillin)
Clonality: monoclonal
Clone: E228
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen