Anti-HDAC2 Rabbit monoclonal antibody unconjugated [clone: Y461]

Lieferant: Abcam
ABCAAB32117-100EA 1020 EUR
ABCAAB32117-100
Anti-HDAC2 Rabbit monoclonal antibody unconjugated [clone: Y461]
Antikörper
Type: Primary
Antigen: HDAC2 (histone deacetylase 2)
Clonality: monoclonal
Clone: Y461
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR