Anti-RAP1GAP Rabbit Monoclonal Antibody [clone: Y135]

Lieferant: Abcam
ABCAAB32119-100EA 688 EUR
ABCAAB32119-100
Anti-RAP1GAP Rabbit Monoclonal Antibody [clone: Y135]
Antikörper
Type: Primary
Antigen: RAP1GAP (RAP1 GTPase activating protein)
Clonality: monoclonal
Clone: Y135
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR