Anti-NCK1 Rabbit Monoclonal Antibody [clone: Y531]

Lieferant: Abcam
ABCAAB32120-100EA 697 EUR
ABCAAB32120-100
Anti-NCK1 Rabbit Monoclonal Antibody [clone: Y531]
Antikörper
Type: Primary
Antigen: NCK1 (NCK adaptor protein 1)
Clonality: monoclonal
Clone: Y531
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR