Anti-RabMAbs None Antibody

Lieferant: Abcam
ABCAAB32121-100EA 896 EUR
ABCAAB32121-100
Anti-RabMAbs None Antibody
Antikörper
Type: Primary
Antigen: RabMAbs
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: None
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR