Anti-BCL2 Rabbit monoclonal antibody unconjugated [clone: E17]

Lieferant: Abcam
ABCAAB32124-100EA 795 EUR
ABCAAB32124-100
Anti-BCL2 Rabbit monoclonal antibody unconjugated [clone: E17]
Antikörper
Type: Primary
Antigen: BCL2 (B-cell CLL/lymphoma 2)
Clonality: monoclonal
Clone: E17
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR