Anti-CRKL Rabbit monoclonal antibody unconjugated [clone: Y243]

Lieferant: Abcam
ABCAAB32126-100EA 688 EUR
ABCAAB32126-100
Anti-CRKL Rabbit monoclonal antibody unconjugated [clone: Y243]
Antikörper
Type: Primary
Antigen: CRKL (v-crk avian sarcoma virus CT10 oncogene homolog-like)
Clonality: monoclonal
Clone: Y243
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR